Learn More
Agilent Technologies PRIMEIT2 RMT PRIMER LABEL KIT
Catalog No. 50125543
Encompass
Labeling Kit; Stratagene; Prime-It RmT; Random Primer Labeling Kit; 25 reactions; Produces high-specific-activity probes in 5 minutes; Stable for 6 months at room temperature; Convenient storage on lab bench and in stockrooms
Catalog No. 50-125-543 Supplier Agilent Technologies Supplier No. 300392
Labeling Kit; Stratagene; Prime-It RmT; Random Primer Labeling Kit; 25 reactions; Produces high-specific-activity probes in 5 minutes; Stable for 6 months at room temperature; Convenient storage on lab bench and in stockrooms