Learn More
Takara Bio PrimeScript™ RT Reagent Kit (Perfect Real Time)
SDP
Catalog No. NC1769765
Encompass
PrimeScript RT Reagent Kit is designed to perform reverse transcription optimized for real-time RT-PCR. It uses PrimeScript Reverse Transcriptase, which features excellent extension. The kit makes fast, efficient cDNA template synthesis for real-time PCR possible. This kit's protocol is simple and suitable for high throughput analysis. It can be used in combination with real-time PCR reagents such as TB Green Premix Ex Taq II (Tli RNaseH Plus) (Cat. #RR820A/B), TB Green Fast qPCR Mix (Cat. #RR430A/B), TB Green Premix Ex Taq (Tli RNaseH Pluse) (Cat. #RR420A/B), or Probe qPCR Mix (Cat. #RR391A/B) for two step real-time RT-PCR. The optimized protocol for assay can be selected in each assay condition using either TB Green or a probe.
Catalog No. NC1769765 Supplier Takara Bio Supplier No. RR037A
PrimeScript RT Reagent Kit is designed to perform reverse transcription optimized for real-time RT-PCR. It uses PrimeScript Reverse Transcriptase, which features excellent extension. The kit makes fast, efficient cDNA template synthesis for real-time PCR possible. This kit's protocol is simple and suitable for high throughput analysis. It can be used in combination with real-time PCR reagents such as TB Green Premix Ex Taq II (Tli RNaseH Plus) (Cat. #RR820A/B), TB Green Fast qPCR Mix (Cat. #RR430A/B), TB Green Premix Ex Taq (Tli RNaseH Pluse) (Cat. #RR420A/B), or Probe qPCR Mix (Cat. #RR391A/B) for two step real-time RT-PCR. The optimized protocol for assay can be selected in each assay condition using either TB Green or a probe.