Learn More
Sielc Technologies Inc PRIMESEP 100 COLUMN
Catalog No. NC9578522
Encompass
Primesep 100 is a reverse-phase analytical column with embedded acidic ion-pairing groups. This is the most versatile column of the entire Primesep family for separation of a broad range of compounds. Improves retention of basic compounds by cation-exchange mechanism. Separates acids by ion-exclusion mechanism. Retains neutral compounds by reverse-phase mechanism. All mobile phases are LC-MS and preparative chromatography compatible. Size: 4.6x150mm
Catalog No. NC9578522 Supplier Sielc Technologies Inc Supplier No. 10046.150.0510
Primesep 100 is a reverse-phase analytical column with embedded acidic ion-pairing groups. This is the most versatile column of the entire Primesep family for separation of a broad range of compounds. Improves retention of basic compounds by cation-exchange mechanism. Separates acids by ion-exclusion mechanism. Retains neutral compounds by reverse-phase mechanism. All mobile phases are LC-MS and preparative chromatography compatible. Size: 4.6x150mm