Learn More
Sielc Technologies Inc Primesep 100 Guard column 4.6xG mm 5um, 100A
Catalog No. NC3665570
Encompass
Primesep 100 is a mixed-mode analytical column with acidic ion-pairing groups embedded into long hydrophobic ligand chains.
Catalog No. NC3665570 Supplier Sielc Technologies Inc Supplier No. 10046.G.0510
Primesep 100 is a mixed-mode analytical column with acidic ion-pairing groups embedded into long hydrophobic ligand chains.