Learn More
Sielc Technologies Inc PRIMESEP 200 HPLC COLUMN,Medium acidic phase
Catalog No. NC9912147
Encompass
Medium acidic phase, Columns with negatively charged functional groups due to the embedded anionic ion-pairing reagent.
Catalog No. NC9912147 Supplier Sielc Technologies Inc Supplier No. 20046.150.0510
Medium acidic phase, Columns with negatively charged functional groups due to the embedded anionic ion-pairing reagent.