Learn More
Sielc Technologies Inc HPLC Column Primesep S2, 100 Å, 5 µm, 4.6 x 150 mm
Catalog No. NC3037070
Encompass
Primesep S2 is a silica-based, acidic column with hydrophilic properties. It provides the following capabilities:– Separates basic compounds using a cation-exchange mechanism.– Separates basic and neutral polar compounds through a HILIC (Hydrophilic Interaction Liquid Chromatography) mechanism.The mobile phases can be tailored for various applications, such as LC-MS and preparative chromatography, and also optimized for low UV detection.
Catalog No. NC3037070 Supplier Sielc Technologies Inc Supplier No. S246.150.0510
Primesep S2 is a silica-based, acidic column with hydrophilic properties. It provides the following capabilities:– Separates basic compounds using a cation-exchange mechanism.– Separates basic and neutral polar compounds through a HILIC (Hydrophilic Interaction Liquid Chromatography) mechanism.The mobile phases can be tailored for various applications, such as LC-MS and preparative chromatography, and also optimized for low UV detection.