Learn More
Takara Bio PRIMESTAR GXL PREMIX 200RXNS
SDP
Catalog No. NC2057968
Encompass
PrimeSTAR GXL is a hot-start, high-fidelity enzyme that is optimized to amplify GC-rich templates, samples with excess templates, and long PCR products up to 30 kb.enables you to simplify your amplification workflow, saving you time and reducing potential errors. Simply add primers and template, and you are ready to go. TAKARA BIO USA INC mfg#R051A
Catalog No. NC2057968 Supplier Takara Bio Supplier No. R051A
PrimeSTAR GXL is a hot-start, high-fidelity enzyme that is optimized to amplify GC-rich templates, samples with excess templates, and long PCR products up to 30 kb.enables you to simplify your amplification workflow, saving you time and reducing potential errors. Simply add primers and template, and you are ready to go. TAKARA BIO USA INC mfg#R051A