Learn More
Takara Bio PRIMESTAR GXL PREMIX,800RXNS
SDP
Catalog No. NC1891833
Encompass
This optimized, 2X master mix is composed of PrimeSTAR GXL DNA Polymerase (a high-fidelity DNA polymerase), reaction buffer, and dNTP mixture. PrimeSTAR GXL Premix is a robust enzyme that can be used in difficult reactions, including amplification of GC-rich regions (without requiring special reaction conditions), in the presence of excess template, and with long targets up to 30 kb in length. Cat. # R051B contains 4 of Cat. # R051A. Please refer to Cat. # R051A for complete product documatentation and resources.
Catalog No. NC1891833 Supplier Takara Bio Supplier No. R051B
This optimized, 2X master mix is composed of PrimeSTAR GXL DNA Polymerase (a high-fidelity DNA polymerase), reaction buffer, and dNTP mixture. PrimeSTAR GXL Premix is a robust enzyme that can be used in difficult reactions, including amplification of GC-rich regions (without requiring special reaction conditions), in the presence of excess template, and with long targets up to 30 kb in length. Cat. # R051B contains 4 of Cat. # R051A. Please refer to Cat. # R051A for complete product documatentation and resources.