Learn More
Takara Bio PrimeSTAR® HS DNA Polymerase (premix), 100 Rxns
SDP
Catalog No. NC0430795
Encompass
A 2X hot-start PCR master mix containing Takara's high-fidelity PrimeSTAR HS DNA polymerase, optimized reaction buffer, and dNTPs. PrimeSTAR HS DNA polymerase has superior proofreading ability due to robust 3' to 5' exonuclease activity. It can efficiently amplify up to 8.5 kb for human genomic DNA targets or up to 22 kb for lambda DNA. Please refer to the User Manual for premix composition. 100 Rxns
Catalog No. NC0430795 Supplier Takara Bio Supplier No. R040A
A 2X hot-start PCR master mix containing Takara's high-fidelity PrimeSTAR HS DNA polymerase, optimized reaction buffer, and dNTPs. PrimeSTAR HS DNA polymerase has superior proofreading ability due to robust 3' to 5' exonuclease activity. It can efficiently amplify up to 8.5 kb for human genomic DNA targets or up to 22 kb for lambda DNA. Please refer to the User Manual for premix composition. 100 Rxns