Learn More
Takara Bio PRIMESTAR HS DNA POL 1000 UNI
SDP
Catalog No. 50443961
Encompass
PCR Enzyme; PrimeSTAR HS DNA Polymerase; 400 rxns; Kit Contents: PrimeSTAR HS DNA Polymerase, 5X PrimeSTAR Buffer (with Mg2sup+), dNTP mixture
Catalog No. 50-443-961 Supplier Takara Bio Supplier No. R010B
PCR Enzyme; PrimeSTAR HS DNA Polymerase; 400 rxns; Kit Contents: PrimeSTAR HS DNA Polymerase, 5X PrimeSTAR Buffer (with Mg2sup+), dNTP mixture