Learn More
Takara Bio PrimeSTAR® HS DNA Polymerase, 250 Units
SDP
Catalog No. 50443960
Encompass
A high-fidelity hot-start (HS) PCR DNA polymerase with superior proofreading ability due to robust 3' to 5' exonuclease activity. PrimeSTAR HS DNA polymerase can efficiently amplify up to 8.5 kb for human genomic DNA targets or up to 22 kb for lambda DNA. Separate tubes of optimzed buffer (Mg2+ plus) and dNTP mix are supplied with the enzyme. 250 Units
Catalog No. 50-443-960 Supplier Takara Bio Supplier No. R010A
A high-fidelity hot-start (HS) PCR DNA polymerase with superior proofreading ability due to robust 3' to 5' exonuclease activity. PrimeSTAR HS DNA polymerase can efficiently amplify up to 8.5 kb for human genomic DNA targets or up to 22 kb for lambda DNA. Separate tubes of optimzed buffer (Mg2+ plus) and dNTP mix are supplied with the enzyme. 250 Units