Learn More
Takara Bio PrimeSTAR® Max DNA Polymerase, 100 Rxns
SDP
Catalog No. NC0421210
Encompass
A 2X hot-start PCR master mix that is ideal for high-fildelity PCR or high-throughput applications. PrimeSTAR Max DNA polymerase pairs Takara Bio's high-fidelity PrimeSTAR HS DNA Polymerase with an elongation factor to provide efficient priming and extension, thereby shortening annealing and extension times. Please refer to the User Manual for the master mix composition. 100 Rxns
Catalog No. NC0421210 Supplier Takara Bio Supplier No. R045A
A 2X hot-start PCR master mix that is ideal for high-fildelity PCR or high-throughput applications. PrimeSTAR Max DNA polymerase pairs Takara Bio's high-fidelity PrimeSTAR HS DNA Polymerase with an elongation factor to provide efficient priming and extension, thereby shortening annealing and extension times. Please refer to the User Manual for the master mix composition. 100 Rxns