Learn More
Takara Bio PRIMESTAR MAX DNA POLY 100RXNS
SDP
Catalog No. NC2957163
Encompass
PrimeSTAR Max DNA Polymerase Ver.2 is a high-performance PCR amplification system that offers the highest levels of fidelity and extension speed. With an improved success rate of target amplification in high-speed reactions compared to PrimeSTAR Max DNA Polymerase (Cat. # R045A/B), PrimeSTAR Max DNA Polymerase Ver.2 maintains the fidelity, making it a fully compatible and superior alternative. As a 2X premixed reagent with an antibody-mediated hot start formulation to suppress DNA Polymerase activity and 3′→5′ exonuclease activity, it allows quick preparation of reactions at room temperature and is useful for high-throughput applications.Reaction size: 1 rxn equals a 50 µl total reaction volume.
Catalog No. NC2957163 Supplier Takara Bio Supplier No. R047A
PrimeSTAR Max DNA Polymerase Ver.2 is a high-performance PCR amplification system that offers the highest levels of fidelity and extension speed. With an improved success rate of target amplification in high-speed reactions compared to PrimeSTAR Max DNA Polymerase (Cat. # R045A/B), PrimeSTAR Max DNA Polymerase Ver.2 maintains the fidelity, making it a fully compatible and superior alternative. As a 2X premixed reagent with an antibody-mediated hot start formulation to suppress DNA Polymerase activity and 3′→5′ exonuclease activity, it allows quick preparation of reactions at room temperature and is useful for high-throughput applications.Reaction size: 1 rxn equals a 50 µl total reaction volume.