Learn More
Longhorn Vaccines And Diagnostics PRIMESTORE ATM 1.5ML RUO 50/PK
SDP
Catalog No. 23100405
Change view
Click to view available options
Product Type:
PrimeStore ATM
Catalog No. Product Type
23-100-405 PrimeStore ATM
1 options
This item is not returnable. View return policy
Catalog No. 23-100-405 Supplier Longhorn Vaccines And Diagnostics Supplier No. LHNATM15
Item Discontinued Please contact a Fisher Healthcare representative for a quote/assistance.
This item is not returnable. View return policy
PrimeStore ATM: An Analyte Transport Medium for optimized molecular testing while preserving and stabilizing proteins and antibodies for rapid testing (no Guanidine or Ethanol).
Specifications
Product Type PrimeStore ATM