Learn More
S BIO PrimeSurface 96 Slit-well Plate
Catalog No. NC1710797
Encompass
PrimeSurface Slit is a newly designed ultra-low attachment 3D plate that improves media exchange handling without disrupting spheroid formation. Cell culture requires frequent media replacement to supply nutrients to growing cells. In conventional 96-well ULA plates, careful aspiration or dispensing is necessary to avoid disturbing unattached spheroids, making the process time-consuming. The PrimeSurface 96 Slit-well Plate allows one-step dispensing or aspiration for all 96 wells simultaneously, reducing pipetting time by over 80% while minimizing spheroid damage risk. This enhancement significantly improves stem cell culture efficiency in regenerative medicine applications.
Catalog No. NC1710797 Supplier S BIO Supplier No. MS9096S
PrimeSurface Slit is a newly designed ultra-low attachment 3D plate that improves media exchange handling without disrupting spheroid formation. Cell culture requires frequent media replacement to supply nutrients to growing cells. In conventional 96-well ULA plates, careful aspiration or dispensing is necessary to avoid disturbing unattached spheroids, making the process time-consuming. The PrimeSurface 96 Slit-well Plate allows one-step dispensing or aspiration for all 96 wells simultaneously, reducing pipetting time by over 80% while minimizing spheroid damage risk. This enhancement significantly improves stem cell culture efficiency in regenerative medicine applications.