Learn More
S BIO Suspension Culture Flask 225 Filter Cap
Catalog No. NC3901756
Encompass
The surface of the S-Bio Suspension Culture Flask is resistant to cell adhesion, and minimize damages and loss of cells when recovering. Ideal for hybridoma and lymphocyte cultivation and useful for LAK adoptive immunity, gene therapy studies, etc. Filter prevents invasion of mycete and bacteria, while permeable to gas.
Catalog No. NC3901756 Supplier S BIO Supplier No. MS2380R
The surface of the S-Bio Suspension Culture Flask is resistant to cell adhesion, and minimize damages and loss of cells when recovering. Ideal for hybridoma and lymphocyte cultivation and useful for LAK adoptive immunity, gene therapy studies, etc. Filter prevents invasion of mycete and bacteria, while permeable to gas.