Learn More
Integrated DNA Technologies Inc PrimeTime Gene Expression Master Mix, High PCR Efficiency >90%, Compatible With Various qPCR Systems, 5 mL
Catalog No. NC1531338
Encompass
The PrimeTime Gene Expression Master Mix is a 2X concentrated solution that enhances accuracy and efficiency in qPCR. It guarantees PCR efficiencies over 90% and R² values above 0.99, making it reliable for gene expression analysis. The mix features exceptional stability at room temperature and high temperatures, reducing shipping costs and environmental impact. Its broad compatibility with various primers and probes differentiates it in the market, perfect for both routine and high-throughput applications.
Catalog No. NC1531338 Supplier Integrated DNA Technologies Inc Supplier No. 1055772
This product needs to be quoted. Please contact your Fisher Scientific Sales Representative for pricing.
The PrimeTime Gene Expression Master Mix is a 2X concentrated solution that enhances accuracy and efficiency in qPCR. It guarantees PCR efficiencies over 90% and R² values above 0.99, making it reliable for gene expression analysis. The mix features exceptional stability at room temperature and high temperatures, reducing shipping costs and environmental impact. Its broad compatibility with various primers and probes differentiates it in the market, perfect for both routine and high-throughput applications.