Learn More
Azer Scientific Primo BP Cassette Printer
SDP
Catalog No. 22026773
Change view
Click to view available options
Color:
White
Catalog No. Color
22-026-773 White
1 options
Catalog No. 22-026-773 Supplier Azer Scientific Supplier No. ES39915BP
Add to Cart
Add to Cart
Primo BP Cassette Printer, batch style printing, includes setup and 2 pre-installed templates. Warranty is only valid with the use of Azer reccomended cassettes.
Azer Scientific's Primo™ batch tissue cassette laser printer is a must-have for pathology and lab environments. Precision construction forges jam-free printing while its low work noise ≤B65 dB design offers ribbon or cartridge-free printing, optimal for cost-efficient workflows.
Specifications
Type Laser Printer
Voltage 100/115/220/230 V
Color White