Learn More
Azer Scientific Primo Cassette Printer
SDP
Catalog No. 22026774
Change view
Click to view available options
Color:
White
Catalog No. Color
22-026-774 White
1 options
Catalog No. 22-026-774 Supplier Azer Scientific Supplier No. ES39915
Add to Cart
Add to Cart
Primo Cassette Printer, includes setup and 2 pre-installed templates. Warranty is only valid with the use of Azer reccomended cassettes.
Easily integrated into laboratory and hospital information systems, this laser cassette printer is perfectly compatible with our hinged tissue cassettes and able to print on cassettes with or without lids. Its structure possesses 6 cassette magazines holding up to 480 cassettes total and the output tray can load 150 cassettes. Choose from many printing options, including 1D as well as 2D barcoding, alphanumeric, graphic, and logo printing. On-demand mode printing, its high printing speed completes a cassette every 3 seconds.
Specifications
Type Laser Printer
Voltage 100/115/220/230 V
Color White