Learn More
Invivogen Primocin- Antimicrobial agent for primary cells, 500 mg (10 x 1 ml)
SDP
Catalog No. NC9141851
Encompass
Unit Size: 500mg (10x1ml vial). Primocin is provided as a sterile light yellow solution at a concentration of 50mg/ml. Sterile water soluble solution. Recommended working concentration: 100ug/ml. One ml Primocin is enough to treat 500ml medium. Primocin is a combination of antibacterial and antifungal compounds. The antibacterial agents eliminate mycoplasma and a wide range of bacteria by blocking both DNA and protein synthesis. The antimycotic agent in Primocin is more stable and less toxic than Amphotericin B. Primocin is non-toxic to primary cells. It acts on targets found only in micro-organisms. The bacterial targets are the DNA gyrase and prokaryotic 30S and 50S ribosomal subunits. The fungal target is ergosterol, a molecule found only in the cell membrane of fungi and yeasts. Shipped at room temperature. For research use only.
Catalog No. NC9141851 Supplier Invivogen Supplier No. ANTPM1
Add to Cart
Add to Cart
Unit Size: 500mg (10x1ml vial). Primocin is provided as a sterile light yellow solution at a concentration of 50mg/ml. Sterile water soluble solution. Recommended working concentration: 100ug/ml. One ml Primocin is enough to treat 500ml medium. Primocin is a combination of antibacterial and antifungal compounds. The antibacterial agents eliminate mycoplasma and a wide range of bacteria by blocking both DNA and protein synthesis. The antimycotic agent in Primocin is more stable and less toxic than Amphotericin B. Primocin is non-toxic to primary cells. It acts on targets found only in micro-organisms. The bacterial targets are the DNA gyrase and prokaryotic 30S and 50S ribosomal subunits. The fungal target is ergosterol, a molecule found only in the cell membrane of fungi and yeasts. Shipped at room temperature. For research use only.