Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Primulin, MP Biomedicals™

Catalog No. ICN19545405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19545405 5 g
ICN19545425 25 g
ICN19545480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19545405 Supplier MP Biomedicals, Inc Supplier No. 0219545405
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8064-60-6
Molecular Formula C21H14N3NaO3S3
Molecular Weight (g/mol) 475.531
MDL Number MFCD00005798
InChI Key RSRNHSYYBLEMOI-UHFFFAOYSA-M
Synonym Direct Yellow 59, C.I. 49000
PubChem CID 3769888
IUPAC Name sodium;2-[2-(4-aminophenyl)-1,3-benzothiazol-6-yl]-6-methyl-1,3-benzothiazole-7-sulfonate
SMILES CC1=C(C2=C(C=C1)N=C(S2)C3=CC4=C(C=C3)N=C(S4)C5=CC=C(C=C5)N)S(=O)(=O)[O-].[Na+]

Specifications

Formula Weight 475.5
Color Brown/Yellow
Quantity 5 g
Physical Form Powder
Chemical Name or Material Primulin
Recommended Storage Store at Room Temperature (15-30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.