Learn More
Andwin Scientific PRIMUS LAB BURNER AND CARTRIDG
SDP
Catalog No. NC0691633
Encompass
The Primus Lab Burner screws into a cartridge to ensure leak-free installation, operation, and removal. The burner also features separate adjustments for regulating gas and air intake.
Catalog No. NC0691633 Supplier Andwin Scientific Supplier No. 470019580
The Primus Lab Burner screws into a cartridge to ensure leak-free installation, operation, and removal. The burner also features separate adjustments for regulating gas and air intake.