Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Principles of PCR with SYBR™ Safe
SDPSCI_ED

Catalog No. 01672269 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
01-672-269 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-269 Supplier EDVOTEK™ Supplier No. 103S
Add to Cart
Add to Cart

In this Principles of PCR kit experiment, students are introduced to the principles and applications of the Polymerase Chain Reaction (PCR). This simulation experiment does not contain human DNA and does not require a thermal cycler.

  • Includes SYBR™ Safe DNA Stain, NOT FlashBlue™ DNA Stain
  • Learn the biology behind polymerase chain reactions (PCR) and gel electrophoresis
  • Explore the relationship between the number of PCR cycles and the quantity of amplified DNA
  • Perform agarose gel electrophoresis to separate different sized DNA molecules
  • Load, run, stain, and analyze DNA samples to observe concentration differences
  • Group Size: For 8 Gels
  • Time Required: Complete in 45 minutes
  • Kit Includes: Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), SYBR™ Safe DNA Stain, Practice Gel Loading Solution.
  • Storage: Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.

Specifications

Grade 8 to Undergraduate
Class Size 8 Lab Groups
Product Type Biotechnology Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.