Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Principles of Thin Layer Chromatography Educational Kit
SDPSCI_ED

Catalog No. S29010 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29010 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29010 Supplier Edvotek™ Supplier No. 113
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The experiment introduces chromatographic theory and methods of thin layer chromatography. Edvotek™ Principles of Thin Layer Chromatography Educational Kit is used for 8 separations.

  • Mixture of dyes are separated on a cellulose-based TLC plate using two different solvent systems
  • Spotting Plates - 20 min; TLC Separation - 5 min
  • Level: Beginner

Recommended Storage

Refrigerator

Specifications

Grade 9 to Undergraduate
Includes Instructions, samples, reagents and solvents, cellulose thin layer plate, 5μL glass capillary pipettes
Quantity 8 Separations
Requires 250mL beakers, metric rulers, pipette pump, 5 or 10mL pipettes, distilled water
Time Range Spotting Plates - 20 min. TLC Separation - 5 min.
Class Size 8 Lab Groups
Product Type Principles of Thin Layer Chromatography Educational Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.