Learn More
Medchemexpress LLC Prinomastat hydrochloride | 1435779-45-5 | 98.2% | 1 ML
SDP
Catalog No. 5000234821
Encompass_Preferred
Prinomastat hydrochloride (AG3340 hydrochloride) is a potent, broad-spectrum matrix metalloproteinase (MMP) inhibitor supplied as a 10 mM solution in DMSO for biochemical and cellular research. It is commonly used to investigate MMP-mediated extracellular matrix remodeling, invasion, and metastasis.
  • Potent, broad-spectrum MMP inhibition for biochemical and cellular assays.
  • Supplied as a ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity, suitable for research applications.
  • Characterized by LC-MS and accompanied by a certificate of analysis.
Catalog No. 50-002-34821 Supplier Medchemexpress LLC Supplier No. HY12170A10MM 1ML
Prinomastat hydrochloride (AG3340 hydrochloride) is a potent, broad-spectrum matrix metalloproteinase (MMP) inhibitor supplied as a 10 mM solution in DMSO for biochemical and cellular research. It is commonly used to investigate MMP-mediated extracellular matrix remodeling, invasion, and metastasis.
  • Potent, broad-spectrum MMP inhibition for biochemical and cellular assays.
  • Supplied as a ready-to-use 10 mM solution in DMSO (1 mL).
  • High purity, suitable for research applications.
  • Characterized by LC-MS and accompanied by a certificate of analysis.