Learn More
GA International Autoclave-Resistant Labels for Inkjet Printers 1.75" x 0.5" / 44.5mm x 12.7mm Rectangle White From -40°C to +150°C (-40°F to +302°F)
Catalog No. NC1493487
Encompass_Preferred
Permanent inkjet printable labels, ideal for glassware, metal and plastic surfaces, trays, and other items intended for use in steam autoclaves, ovens, and gamma irradiators (tested up to 50 KGy). Our autoclave inkjet printable labels are durable, weather & water-proof, alcohol-resistant, and have been tested to endure 10 autoclave cycles (up to 121°C/250°F, 16.5 psi for 20 minutes). Compatible with all major brands of inkjet printers and are easy to format for use with our free online Microsoft Word templates.
Catalog No. NC1493487 Supplier GA International Supplier No. INKA13WH
Permanent inkjet printable labels, ideal for glassware, metal and plastic surfaces, trays, and other items intended for use in steam autoclaves, ovens, and gamma irradiators (tested up to 50 KGy). Our autoclave inkjet printable labels are durable, weather & water-proof, alcohol-resistant, and have been tested to endure 10 autoclave cycles (up to 121°C/250°F, 16.5 psi for 20 minutes). Compatible with all major brands of inkjet printers and are easy to format for use with our free online Microsoft Word templates.