Learn More
LGI PRINTED QUARANTINE DOT LABEL
Supplier:  LGI 66133
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Removable 2x2 red dot labels from Lgi. ISO 9001-2000 certified. RoHS compliant. 1k labels/roll. Min. order: 2 rolls. Lgi also creates innovative custom labels that meet your specific cleanroom requirements. Based on their proprietary CleanMark process, engineers and designers will work with you to design and construct labels to fit any size, permanent or removable adhesive, direct thermal or thermal transfer printable, can withstand extreme temperatures, harsh chemicals and abrasion. Labels can indicate sterilization such as autoclave or gamma or can be pre-sterilized. 100% synthetic, non-particulating, ultra low-outgassing raw materials wont contaminate clean or sterile environments. Lgis exclusive EZ-Peel feature is adhesive-free on one or both sides of the label, making it easy to apply and remove with protective gloves typical in cleanrooms, fabs, medical labs and other controlled environments.
Catalog No. 50-112-6984