Learn More
Perkin Elmer US LLC Printer Cable - Centronics, Parallel
Catalog No. 502112925
Encompass_Preferred
The Centronics Parallel Printer Cable is 6 feet long. One end has a DB5 male connector with thumb screws on a 25 pin D shell. The opposite end screws into a Centronics 36 Male connector.
Catalog No. 50-211-2925 Supplier Perkin Elmer US LLC Supplier No. 09410057
The Centronics Parallel Printer Cable is 6 feet long. One end has a DB5 male connector with thumb screws on a 25 pin D shell. The opposite end screws into a Centronics 36 Male connector.