Learn More
TUTTNAUER USA PRINTER F/LABSCI 60LV AND 85LV
Supplier:  TUTTNAUER USA THE0020116
Encompass_Preferred
Thermal printer for LabSci 60LV & 85LV. Printer is capable of printing barcode labels or sterilizer cycle information. Need to order thermal paper rolls or thermal barcode label rolls depending on customer application.
Catalog No. NC2542684