Learn More
Research Products International Corp Printer, Impact, SF40A, AM
SDP
Supplier:  Research Products International Corp 30064203
Encompass
The SF40A combines high performance and multiple functions to offer OHAUS quality and durability in a portable printer. The SF40A offers advanced functionality in an easy-to-use and set up printer at an exceptional value. This versatile printer is ideal for use in conjunction with OHAUS balances and scales in laboratory, industrial, and educational applications to ensure accurate documentation of weighing results.
The statistics and totalization modes and real time clock complement OHAUS weighing instruments’ application modes and offer functionality that assists the reporting processes.
SF40A helps support the efficiency of the weighing and recording processes. RS232 interface and PC configuration software make setting up and using the printer fast and simple.
With the ability to print in 13 different languages, SF40A can print in the language of the connected device to support universal operation.
Catalog No. 50-197-6953