Learn More
Epredia™ PrintMate™ AS Cassette Printer Total Care Package
Catalog No. A83800001B Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
PrintMate AS 150 Cassette Printer
PrintMate AS 150 Cassette Printing System
PrintMate AS 450 Cassette Printer
PrintMate AS 450 Cassette Printing System
PrintMate AS 900 Cassette Printer
PrintMate AS 900 Cassette Printing System
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
A83800001B PrintMate AS 150 Cassette Printer
A83800004B PrintMate AS 150 Cassette Printing System
A83800002B PrintMate AS 450 Cassette Printer
A83800005B PrintMate AS 450 Cassette Printing System
A83800003B PrintMate AS 900 Cassette Printer
A83800006B PrintMate AS 900 Cassette Printing System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. A83800001B Supplier Epredia™ Supplier No. A83800001B
Add to Cart
Add to Cart
Request a Quote
The PrintMate AS Total Care Package offers a complete solution, combining the PrintMate AS on-demand cassette printer with cassettes and total care service support.
The PrintMate AS Total Care Package offers workflow protection by offering a complete solution, combining the PrintMate AS on-demand cassette printer with cassettes and total care service support.
Cassettes
  • The entire range of cassettes has been redesigned for better performance – both in printing and in laboratory processes.
  • The all-new surface finish and reinforced print faces are built to optimize print quality when used with PrintMate AS cassette printers.
  • All-new PrintMate AS-compatible tubes are precisely engineered to reduce jamming and prevent downtime.
  • Opening and closing forces have been carefully adjusted for ease of use, while safeguarding valuable specimens during processing.
  • All cassette designs have been comprehensively validated for print quality, processing quality, and reliability in embedding and sectioning.
  • New 12-color range selected for scanning reliability.
Total Care Service Support
  • The PrintMate AS Total Care Package includes three years of service support – one year warranty and two years extended service.
  • Total Care Package customers receive two preventative visits per year for three years.
  • At these visits, wear components are automatically replaced to ensure reliable operation and uninterrupted workflow.
Specifications
Description PrintMate AS 150 Cassette Printer Plus Extended Two Years of Service Support
For Use With (Equipment) PrintMate AS 150 Cassette Printer
Type PrintMate AS Total Care Package