Learn More
Epredia™ PrintMate™ Biopsy Cassettes with Lids in Pre-packed Tubes, aqua
Catalog No. A84210111 Shop All Epredia Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
A84210111 Blue
1 options
This item is not returnable. View return policy
Catalog No. A84210111 Supplier Epredia™ Supplier No. A84210111
This item is not returnable. View return policy
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. Packed in tubes, PrintMate Cassettes are intended for high throughput printing, and provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
PrintMate Biopsy Cassettes in pre-packed tubes. Embedding Casette Tubes. 40 Tubes X 75 Cassettes, total 3000 Cassettes with 3000 Lids.
Specifications
Pore Style Small Square Pore
PrintMate Compatible Yes
Color Blue
Product Type Biopsy Cassette