Learn More
Epredia™ PrintMate™ Embedding Cassettes with Lids, aqua
Catalog No. A84210076 Shop All Epredia Products
Change view
Click to view available options
Color:
Green
Pink
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
A84210076 Pink
A84210072 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. A84210076 Supplier Epredia™ Supplier No. A84210076
This item is not returnable. View return policy
Epredia™ PrintMate Cassettes™ are designed to contain incised tissue safely and securely for tissue processing, embedding and sectioning. PrintMate Cassettes provide exceptional scanning accuracy and reliability when used with the Epredia™ PrintMate™ and Epredia™ SlideMate™ barcode scanner.
Specifications
Pore Style Round Pore
PrintMate Compatible Yes
Color Pink
Product Type Embedding Biopsy Cassette