Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ 3 in. BLUE MSDS BINDER

Catalog No. 19103237 Shop All Brady Corporation Products
Catalog No. 19-103-237 Supplier Brady™ Supplier No. BR755B
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Package includes binder only (information center and accessories sold separately)

3 in. BLUE MSDS

Specifications

Format Binder
Includes Binder
Diameter (English) Ring 3 in.
Height (English) 11 in.
Quantity Each
Depth (English) 3.5 in.
Languages English
For Use With (Application) Hazard Communication
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.