Description
- For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps
- Reduces the number of locks being used
- Use the SAFETY REDBOX™ Lock Box as a portable or mounted lockbox.
- Quick release internal slide button allows lockbox to be carried to the poind of need
Specifications
Specifications
| Product Type | Electrical and Mechanical Risk Lockouts |
| Color | Gray |
| Includes | (10) circuit breaker lockouts; (1) cable lockout with cable; (3) yellow lockout hasp; (3) label; (1) booklet; (3) padlock; (2) valve lockout; (1) box |
| Quantity | 1 Kit |