Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ ECON RTK CENTER, WITH INSERTS

Catalog No. 19103273 Shop All Brady Corporation Products
Change view
Click to view available options
For Use With (Application):
Hazard Communication
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
19-103-273 Hazard Communication
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-103-273 Supplier Brady™ Supplier No. 102720

missing translation for 'validMainframeMsg'

Safety information centers provide a centralized location for important safety information and makes the information accessible to employees.

  • Helps prevent costly mistakes by providing potentially life-saving information to employees
  • Ideal for improving awareness of your company's safety information

Specifications

Format Wall Mounted
Product Type Information Center
Quantity Kit
For Use With (Application) Hazard Communication
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.