Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ MSDS 1-1/2 INCH BLUE BINDER

Catalog No. 19103223 Shop All Brady Corporation Products
Catalog No. 19-103-223 Supplier Brady™ Supplier No. BR361B
  • $136.00
    $61.65 / Each of 1

    Save $74.35

    55% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Package includes binder only (information center and accessories sold separately)

MSDS 1-1/2 INCH BLUE BINDER

Specifications

Format Binder
Includes Binder; Inserts
Diameter (English) Ring 1.5 in.
Height (English) 11 in.
Quantity Each
Depth (English) 2 in.
Material Polyethylene
Color Yellow on Blue
Languages English
For Use With (Application) Hazard Communication
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.