Learn More
Enzo Life Sciences Prion protein monoclonal antibody (3F4) (200µg)
SDP
Catalog No. 502012678
Encompass_Preferred
Clone: 3F4. Isotype: IgG2a. Species reactivity: Human Hamster, Cat. Applications: ELISA, IHC, IP, WB. Purity: Purified. Formulation: Liquid. In PBS.
Catalog No. 50-201-2678 Supplier Enzo Life Sciences Supplier No. ENZABS1190200
Clone: 3F4. Isotype: IgG2a. Species reactivity: Human Hamster, Cat. Applications: ELISA, IHC, IP, WB. Purity: Purified. Formulation: Liquid. In PBS.