Learn More
Research Products International Corp Prism Refrigerated Micro Centrifuge
SDP
Catalog No. 501976808
Encompass
The Prism R Refrigerated Micro-Centrifuge features the most efficient cooling system available for a benchtop instrument. Achieves 4°C in just eight minutes and maintains temperature at maximum speed. Powerful brushless motor provides high speed up to 13,500 rpm and is maintenance free. Includes a 24 position solid aluminum rotor designed for easy access to tubes. One ergonomic knob controls selection of time, speed with Quick-Cool option. View actual speed, temperature and remaining spin time on a large LCD panel. This extremely quiet centrifuge produces a low noise level of 56db.
Designed for reliable and maintenance free performance and includes a two year warranty.
Catalog No. 50-197-6808 Supplier Research Products International Corp Supplier No. 141410
Add to Cart
Add to Cart
Request a Quote
The Prism R Refrigerated Micro-Centrifuge features the most efficient cooling system available for a benchtop instrument. Achieves 4°C in just eight minutes and maintains temperature at maximum speed. Powerful brushless motor provides high speed up to 13,500 rpm and is maintenance free. Includes a 24 position solid aluminum rotor designed for easy access to tubes. One ergonomic knob controls selection of time, speed with Quick-Cool option. View actual speed, temperature and remaining spin time on a large LCD panel. This extremely quiet centrifuge produces a low noise level of 56db.
Designed for reliable and maintenance free performance and includes a two year warranty.