Learn More
Medchemexpress LLC Pristane-d40 | 16416-35-6 | 10 MG
SDP
Catalog No. 5000418853
Encompass_Preferred
Pristane-d40 is the deuterium labeled Pristane, a naturally occurring hydrocarbon oil. It is found in small quantities in many plants, various marine organisms, and as the most active component of mineral oil. This product is a non-antigenic adjuvant that induces MHC class II-restricted, arthritogenic T cells in rats.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-18853 Supplier Medchemexpress LLC Supplier No. HYN7819S10MG
Pristane-d40 is the deuterium labeled Pristane, a naturally occurring hydrocarbon oil. It is found in small quantities in many plants, various marine organisms, and as the most active component of mineral oil. This product is a non-antigenic adjuvant that induces MHC class II-restricted, arthritogenic T cells in rats.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS