Learn More
Medchemexpress LLC Pristane-d40 | 16416-35-6 | 25 MG
SDP
Catalog No. 5000421841
Encompass_Preferred
Pristane-d40 is the deuterium labeled Pristane. Pristane (Norphytane) is a naturally occurring hydrocarbon oil found in small quantities in many plants, various marine organisms, and as the most active component of mineral oil. It is a non-antigenic adjuvant and induces MHC class II-restricted, arthritogenic T cells in the rat.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-21841 Supplier Medchemexpress LLC Supplier No. 25MGHYN7819S
Pristane-d40 is the deuterium labeled Pristane. Pristane (Norphytane) is a naturally occurring hydrocarbon oil found in small quantities in many plants, various marine organisms, and as the most active component of mineral oil. It is a non-antigenic adjuvant and induces MHC class II-restricted, arthritogenic T cells in the rat.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.