Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pristane-d40 | 16416-35-6 | 5 MG
SDP

Catalog No. 5000417324
Encompass_Preferred

Pristane-d40 is a deuterium-labeled version of Pristane (Norphytane), a naturally occurring hydrocarbon oil found in plants, various marine organisms, and as an active component of mineral oil. It functions as a non-antigenic adjuvant, inducing MHC class II-restricted, arthritogenic T cells in rats. Deuteration, which involves substituting hydrogen with deuterium, can affect the pharmacokinetic and metabolic profiles of drugs.

  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular weight of 308.77
  • Chemical formula of C19D40
  • Appearance is a colorless to light yellow liquid
  • Purity of 98.56%

Catalog No. 50-004-17324 Supplier Medchemexpress LLC Supplier No. HYN7819S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pristane-d40 is a deuterium-labeled version of Pristane (Norphytane), a naturally occurring hydrocarbon oil found in plants, various marine organisms, and as an active component of mineral oil. It functions as a non-antigenic adjuvant, inducing MHC class II-restricted, arthritogenic T cells in rats. Deuteration, which involves substituting hydrogen with deuterium, can affect the pharmacokinetic and metabolic profiles of drugs.

  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Molecular weight of 308.77
  • Chemical formula of C19D40
  • Appearance is a colorless to light yellow liquid
  • Purity of 98.56%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.