Learn More
TARGETMOL CHEMICALS INC PRISTIMERIN 10MG
SDP
Catalog No. 502576718
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. 1. Pristimerin (Celastrol methyl ester) has anticancer activity treatment with Pristimerin may be a potential strategy for the suppression of hypoxia-induced metastasis through the reversal of hypoxia-induced stem cell characteristics and EMT in cancer cells. 2. Pristimerin down-regulates the expression of pro-inflammatory mediators through blocking of NF-(kappa)B activation by inhibiting interconnected ROS/IKK/NF-(kappa)B signaling pathways can effectively inhibit both arthritic inflammation and cartilage and bone damage in the joints. purity: 99%
Catalog No. 50-257-6718 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S105010MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. 1. Pristimerin (Celastrol methyl ester) has anticancer activity treatment with Pristimerin may be a potential strategy for the suppression of hypoxia-induced metastasis through the reversal of hypoxia-induced stem cell characteristics and EMT in cancer cells. 2. Pristimerin down-regulates the expression of pro-inflammatory mediators through blocking of NF-(kappa)B activation by inhibiting interconnected ROS/IKK/NF-(kappa)B signaling pathways can effectively inhibit both arthritic inflammation and cartilage and bone damage in the joints. purity: 99%