Learn More
Medchemexpress LLC HY-N1937 10mg Medchemexpress, Pristimerin CAS:1258-84-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN193710MG
Encompass_Preferred
Medchemexpress, HY-N1937 10mg Pristimerin CAS:1258-84-0 Pristimerin is a potent and reversible monoacylglycerol lipase ( MGL ) inhibitor with an IC 50 of 93 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3860