Learn More
Medchemexpress LLC Pristimerin | 1258-84-0 | 464.64 | 5 MG
SDP
Catalog No. 5000416642
Encompass_Preferred
Pristimerin is a potent and reversible monoacylglycerol lipase (MGL) inhibitor with an IC50 of 93 nM. It is also known as Celastrol methyl ester. This compound is used for research purposes.
  • Potent and reversible monoacylglycerol lipase (MGL) inhibitor.
  • Inhibits angiogenesis by suppressing VEGFR2 signaling pathways.
  • Exhibits anti-inflammatory properties.
  • Demonstrates anti-tumorigenic properties and cytotoxicity against various human cancer cell lines.
  • Attenuates activation of the VEGF-induced VEGFR2-mediated signaling pathway.
  • Reduces vessel density in synovial membrane tissues.
  • Decreases expression of pro-angiogenic factors like TNF-α, Ang-1, and MMP-9 in sera.
Catalog No. 50-004-16642 Supplier Medchemexpress LLC Supplier No. HYN19375MG
Pristimerin is a potent and reversible monoacylglycerol lipase (MGL) inhibitor with an IC50 of 93 nM. It is also known as Celastrol methyl ester. This compound is used for research purposes.
  • Potent and reversible monoacylglycerol lipase (MGL) inhibitor.
  • Inhibits angiogenesis by suppressing VEGFR2 signaling pathways.
  • Exhibits anti-inflammatory properties.
  • Demonstrates anti-tumorigenic properties and cytotoxicity against various human cancer cell lines.
  • Attenuates activation of the VEGF-induced VEGFR2-mediated signaling pathway.
  • Reduces vessel density in synovial membrane tissues.
  • Decreases expression of pro-angiogenic factors like TNF-α, Ang-1, and MMP-9 in sera.