Learn More
Cayman Chemical Pristimerin, Purity: ≥98%, CAS: 1258-84-0, 50 mg
SDP
Catalog No. 502953504
Encompass_Preferred
A naturally occuring terpenoid that potently inhibits MAGL (IC50 = 93 nM); at 1 µM, significantly inhibits endogenous MAGL in isolated rat neurons
Catalog No. 50-295-3504 Supplier Cayman Chemical Supplier No. 1362150 MG
A naturally occuring terpenoid that potently inhibits MAGL (IC50 = 93 nM); at 1 µM, significantly inhibits endogenous MAGL in isolated rat neurons