Learn More
BIRCH BIOTECH LLC PRISTINE Acetonitrile, HPLC Grade, 4 Liter, 4/case
Supplier:  BIRCH BIOTECH LLC 19105
Encompass_Preferred
PRISTINE Acetonitrile, HPLC Grade, 4 Liter, 4/casePrecision Solvent for High-Performance Liquid Chromatography (HPLC)PRISTINE HPLC Grade Acetonitrile is a high-purity solvent specially formulated for high-performance liquid chromatography applications. With a purity level of ≥99.9%, it’s ideal for accurate and reliable analytical results in both research and industrial settings. Engineered to deliver consistent results, our HPLC-grade acetonitrile minimizes background noise and reduces contamination risks, enabling high sensitivity and precision in analytical detection.
Catalog No. NC3861138