Learn More
BIRCH BIOTECH LLC PRISTINE Acetonitrile, LC-MS Grade, 4 Liter, 4/case
Catalog No. NC3861136
Encompass_Preferred
PRISTINE Acetonitrile, LC-MS Grade, 4 Liter, 4/caseUltra-Pure Solvent for Liquid Chromatography-Mass Spectrometry (LC-MS)PRISTINE LC-MS Grade Acetonitrile is a premium, ultra-pure solvent meticulously designed for liquid chromatography-mass spectrometry applications, offering unmatched performance and reliability for the most demanding analytical procedures. With a purity of ≥99.99% and an exceptionally low level of metal ions and organic impurities, this acetonitrile grade is ideal for sensitive MS detection, delivering accurate and reproducible results for both qualitative and quantitative analyses.
Catalog No. NC3861136 Supplier BIRCH BIOTECH LLC Supplier No. 19305
PRISTINE Acetonitrile, LC-MS Grade, 4 Liter, 4/caseUltra-Pure Solvent for Liquid Chromatography-Mass Spectrometry (LC-MS)PRISTINE LC-MS Grade Acetonitrile is a premium, ultra-pure solvent meticulously designed for liquid chromatography-mass spectrometry applications, offering unmatched performance and reliability for the most demanding analytical procedures. With a purity of ≥99.99% and an exceptionally low level of metal ions and organic impurities, this acetonitrile grade is ideal for sensitive MS detection, delivering accurate and reproducible results for both qualitative and quantitative analyses.