Learn More
BIRCH BIOTECH LLC PRISTINE Dichloromethane, HPLC Grade, 4 Liter, 4/case
Catalog No. NC3861157
Encompass_Preferred
PRISTINE Dichloromethane, HPLC Grade, 4 Liter, 4/caseHigh-Purity Solvent for High-Performance Liquid Chromatography (HPLC)PRISTINE HPLC Grade Dichloromethane is a high-purity solvent designed to meet the stringent requirements of high-performance liquid chromatography and other sensitive analytical techniques. With exceptional purity and low UV absorbance, this dichloromethane grade minimizes background interference, enabling precise, reliable results in both qualitative and quantitative analyses. Its stability and consistent performance make it ideal for demanding applications across pharmaceutical, environmental, and biotech industries.
Catalog No. NC3861157 Supplier BIRCH BIOTECH LLC Supplier No. 19135
PRISTINE Dichloromethane, HPLC Grade, 4 Liter, 4/caseHigh-Purity Solvent for High-Performance Liquid Chromatography (HPLC)PRISTINE HPLC Grade Dichloromethane is a high-purity solvent designed to meet the stringent requirements of high-performance liquid chromatography and other sensitive analytical techniques. With exceptional purity and low UV absorbance, this dichloromethane grade minimizes background interference, enabling precise, reliable results in both qualitative and quantitative analyses. Its stability and consistent performance make it ideal for demanding applications across pharmaceutical, environmental, and biotech industries.