Learn More
BIRCH BIOTECH LLC PRISTINE Isopropyl Alcohol, Electronic Grade, 4x1 gallon per case
Catalog No. NC3861160
Encompass_Preferred
PRISTINE Isopropyl Alcohol, Electronic Grade, 4x1 gallon per caseHigh-Purity Solvent for Electronic Grade Applications, PRISTINE Electronic Grade Isopropyl Alcohol, is used to cleanse organic matters or remove water/moisture of the surface. It also is a rgeat buffer solution to prevent anti-corrosion for wiring.
Catalog No. NC3861160 Supplier BIRCH BIOTECH LLC Supplier No. 19855
PRISTINE Isopropyl Alcohol, Electronic Grade, 4x1 gallon per caseHigh-Purity Solvent for Electronic Grade Applications, PRISTINE Electronic Grade Isopropyl Alcohol, is used to cleanse organic matters or remove water/moisture of the surface. It also is a rgeat buffer solution to prevent anti-corrosion for wiring.